Z568171
Aldrich® filtering flask, heavy-wall
capacity 250 mL, neck joint: ST/NS 24/40
About This Item
Productos recomendados
material
borosilicate glass
joint
joint: ST/NS neck24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
250 mL
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC