Key Documents
Z551708
Chemat vacuum chucks
diam. 2 in.
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
description
Type CG
manufacturer/tradename
Chemat
diam.
2 in.
General description
For use with the Chemat Spin-coater system.