Key Documents
SYNH200003
Synthware aluminum heat transfer block for round bottom flasks
About This Item
UNSPSC Code:
41122800
NACRES:
NB.51
manufacturer/tradename
H200003
General description
holds 5, 10 and 25mL round bottomflasks, one hold on the top for a standard thermometer.
Legal Information
Synthware is a trademark of Kemtech America Inc.