HPLC Analysis of 13 Aldehyde/Ketone DNPH Derivatives on Ascentis® Express C18
Materials
analytical column
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express C18, 10 cm x 4.6 mm I.D., 2.7 μm particles (53827-U)
mobile phase
[A] water; [B] acetonitrile; (40:60 v/v, A:B)
flow rate
2 mL/min
pressure
4685 psi (323 bar)
detector
UV, 360 nm
injection
10 μL
sample
carb Method 1004 DNPH Mix 2 (47651-U) dilute 3:2 with water
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany