Merck
  • Synergistic coordination of polyethylene glycol with ClpB/DnaKJE bichaperone for refolding of heat-denatured malate dehydrogenase.
Synergistic coordination of polyethylene glycol with ClpB/DnaKJE bichaperone for refolding of heat-denatured malate dehydrogenase.
Biotechnology progress (2009-06-25)
Rui Nian, Duck Sang Kim, Lihan Tan, Chan-Wha Kim, Woo-Seok Choe
ABSTRACT
The use of polyethylene glycol (PEG) as a refolding additive to a refolding cocktail comprising the molecular bichaperone ClpB and DnaKJE significantly enhances chaperone-mediated refolding of heat-denatured malate dehydrogenase (MDH). The critical factor to affect the refolding yield is the time point of introducing PEG to the refolding cocktail. The refolding efficiency reached approximately 90% only when PEG was added at the beginning of refolding reaction. The synergistic coordination of an inexpensive refolding additive PEG with the ClpB/DnaKJE bichaperone system may provide an economical route to further enhance the efficacy of ClpB/DnaKJE refolding cocktail approach, facilitating its implementation in large-scale refolding processes.
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, Grade I, ≥99%, from microbial
Pricing and availability is not currently available.