Merck
  • Modulation of the main porcine enteric neuropeptides by a single low-dose of lipopolysaccharide (LPS)
Modulation of the main porcine enteric neuropeptides by a single low-dose of lipopolysaccharide (LPS)
Gut pathogens (2017-12-20)
Anita Mikołajczyk, Sławomir Gonkowski, Dagmara Złotkowska
ABSTRACT
The present research was conducted to investigate the influence of a low, single dose of LPS, which does not result in any clinical symptoms of intoxication on the expression of selected neuropeptides within the intestines of the domestic pig. This experiment was conducted on immature female pigs of the Pitrain × Duroc breed (n = five per group). Seven days after the intravenous injection of 10 mL saline solution for control animals and 5 μg/kg b.w. (in 10 mL saline solution) LPS The results of this research indicate that single low-dose LPS The exact functions of the substances studied and mechanisms of responses to LPS action depend on the sections of the intestines. The mechanisms of observed changes are not fully understood, but fluctuations in neuronal active substance levels may be connected with neurodegenerative and/or pro-inflammatory activity of LPS.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype enteritidis, γ-irradiated, BioXtra, suitable for cell culture
Pricing and availability is not currently available.