Merck
  • MST4 Phosphorylation of ATG4B Regulates Autophagic Activity, Tumorigenicity, and Radioresistance in Glioblastoma.
MST4 Phosphorylation of ATG4B Regulates Autophagic Activity, Tumorigenicity, and Radioresistance in Glioblastoma.
Cancer cell (2017-12-13)
Tianzhi Huang, Chung Kwon Kim, Angel A Alvarez, Rajendra P Pangeni, Xuechao Wan, Xiao Song, Taiping Shi, Yongyong Yang, Namratha Sastry, Craig M Horbinski, Songjian Lu, Roger Stupp, John A Kessler, Ryo Nishikawa, Ichiro Nakano, Erik P Sulman, Xinghua Lu, Charles David James, Xiao-Ming Yin, Bo Hu, Shi-Yuan Cheng
ABSTRACT
ATG4B stimulates autophagy by promoting autophagosome formation through reversible modification of ATG8. We identify ATG4B as a substrate of mammalian sterile20-like kinase (STK) 26/MST4. MST4 phosphorylates ATG4B at serine residue 383, which stimulates ATG4B activity and increases autophagic flux. Inhibition of MST4 or ATG4B activities using genetic approaches or an inhibitor of ATG4B suppresses autophagy and the tumorigenicity of glioblastoma (GBM) cells. Furthermore, radiation induces MST4 expression, ATG4B phosphorylation, and autophagy. Inhibiting ATG4B in combination with radiotherapy in treating mice with intracranial GBM xenograft markedly slows tumor growth and provides a significant survival benefit. Our work describes an MST4-ATG4B signaling axis that influences GBM autophagy and malignancy, and whose therapeutic targeting enhances the anti-tumor effects of radiotherapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG7
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG5
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG4B
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit