Merck
  • Epoxyazadiradione Purified from the Azadirachta indica Seed Induced Mitochondrial Apoptosis and Inhibition of NFκB Nuclear Translocation in Human Cervical Cancer Cells.
Epoxyazadiradione Purified from the Azadirachta indica Seed Induced Mitochondrial Apoptosis and Inhibition of NFκB Nuclear Translocation in Human Cervical Cancer Cells.
Phytotherapy research : PTR (2017-10-19)
G Shilpa, J Renjitha, R Saranga, Francis K Sajin, Mangalam S Nair, Beena Joy, B S Sasidhar, S Priya
ABSTRACT
Epoxyazadiradione (EAD) is an important limonoid present in Neem (Azadirachta indica) plant. In the present study, we have purified EAD from Neem seed and studied its anticancer potential in human cervical cancer (HeLa) cells. Cell proliferation inhibition studies indicated that the GI
Sigma-Aldrich
Human CCL21 / 6ckine ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)