Merck
  • Anodization of magnesium for biomedical applications - Processing, characterization, degradation and cytocompatibility.
Anodization of magnesium for biomedical applications - Processing, characterization, degradation and cytocompatibility.
Acta biomaterialia (2017-08-19)
Aaron F Cipriano, Jiajia Lin, Christopher Miller, Alan Lin, Mayra C Cortez Alcaraz, Pedro Soria, Huinan Liu
ABSTRACT
This article reports anodization of Mg in KOH electrolyte and the associated surface, degradation, and biological properties for bioresorbable implant applications. The preparation procedures for electrodes and anodization setup significantly enhanced reproducibility of samples. The results of anodization performed at the applied potentials of 1.8, 1.9, or 2.0V showed that the sample anodized at 1.9V and annealed, referred to as the 1.9 AA sample, had homogenous surface microstructure and elemental composition, and a reduction in corrosion current density in the electrochemical testing. In comparison with Mg control, the 1.9 AA sample showed a distinct mode of degradation, e.g., continuous growth of a passivation layer enriched with Ca and P instead of typical localized pitting and undermining, and a greater release rate of Mg Magnesium (Mg)-based biomaterials have been specifically designed and actively explored for biodegradable implant applications since the early 2000s. To realize the benefits of Mg-based materials for medical implant applications, it is critical to control the rate of Mg degradation (i.e. corrosion) in the body. We investigated an environmentally friendly anodization process using KOH electrolyte for modifying the surface of Mg-based materials, and the resulted surface, degradation, and biological properties for biomedical applications. This study reported critical considerations that are important for repeatability of anodization process, homogeneity of surface microstructure and composition, and in vitro evaluations of the degradation and biological properties of surface treated Mg samples. The details in preparation of electrodes, anodization setup, annealing, and sample handling before and after surface treatment (e.g. re-embedding) reported in this article are valuable for studying a variety of electrochemical processes for surface treatment of Mg-based metals, because of enhanced reproducibility.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
SKU
Pack Size
Availability
Price
Quantity
Nalgene® Griffin beakers, capacity 50 mL, translucent polypropylene