Merck
  • The high mobility group AT-hook 1 protein stimulates bovine herpesvirus 1 productive infection.
The high mobility group AT-hook 1 protein stimulates bovine herpesvirus 1 productive infection.
Virus research (2017-07-08)
Liqian Zhu, Clinton Jones
ABSTRACT
Bovine herpesvirus 1 (BoHV-1) is an important pathogen of cattle that causes clinical symptoms in the upper respiratory tract and conjunctivitis. Like most alpha-herpesvirinae subfamily members, BoHV-1 establishes latency in sensory neurons. Stress consistently induces reactivation from latency, which is essential for virus transmission. Recent studies demonstrated that a viral protein (ORF2) expressed in a subset of latently infected neurons is associated with β-catenin and the high mobility group AT-hook 1 protein (HMGA1), which correlates with increased expression of these proteins in latently infected neurons. Since HMGA1 is primarily expressed in actively growing cells, binds to the minor groove of A+T rich regions in double-stranded DNA, and mediates gene transcription, we hypothesized that HMGA1 regulates BoHV-1 productive infection. Studies in this report indicated that productive infection increased HMGA1 protein levels and re-localized the protein in the nucleus. Netropsin, a small molecule that binds to the minor groove of DNA and prevents HMGA1 from interacting with DNA inhibited viral replication and interfered with the ability of BoHV-1 to induce HMGA1 re-localization. Furthermore, netropsin reduced RNA and protein expression of two viral regulatory proteins (bICP0 and bICP22) during productive infection, but increased bICP4 levels. Small interfering RNAs (siRNAs) that specifically target HMGA1 reduced HMGA1 RNA levels and virus production confirming HMGA1 stimulates productive infection.
Sigma-Aldrich
MISSION® esiRNA, targeting human HMGA1
Pricing and availability is not currently available.