Merck
  • Inhibitory modulation of human estrogen receptor α and β activities by dicyclohexyl phthalate in human breast cancer cell lines.
Inhibitory modulation of human estrogen receptor α and β activities by dicyclohexyl phthalate in human breast cancer cell lines.
The Journal of toxicological sciences (2017-07-19)
Hiroyuki Okazaki, Shuso Takeda, Saki Matsuo, Masaya Matsumoto, Erika Furuta, Eriko Kohro-Ikeda, Hironori Aramaki
ABSTRACT
Phthalate esters (PAEs) are man-made compounds that are used widely in industry, and the ubiquitous exposure of humans to PAEs has been reported. Although some PAEs have been suggested to function as xenoestrogens in in vitro systems, such as human estrogen receptors (ERs) expressed in Chinese hamster ovary (CHO)-K1 cells, few studies have attempted to elucidate whether PAEs affect human ERα/ERβ-mediated signaling in human breast cancer cells (i.e., combination between human ERs and human cells). Thus, further experiments are needed in order to clarify the activities of PAEs. Among the 9 PAEs (carbon# in the side chains: 2-8) investigated, dibutyl phthalate (DBP), dipentyl phthalate (DPENP), and dicyclohexyl phthalate (DCHP) were found to exhibit strong anti-estrogenic activities in MCF-7 cells (ER-positive) in the presence of 1 nM 17β-estradiol (E2). Since limited information is currently available on DPENP and DCHP, we herein focused on these two PAEs. Experiments using MDA-MB-231 cells (ER-negative) transfected with human ERα or ERβ expression plasmids revealed that DCHP was a markedly stronger anti-estrogenic PAE than DPENP; DCHP inhibited ERα and ERβ activities stimulated by 1 nM E2 with IC
Sigma-Aldrich
2,3-Bis(4-hydroxyphenyl)propionitrile, ≥98% (HPLC)
Supelco
Dihexyl phthalate, analytical standard