Merck
Identification of Buctopamine and Mebuctopamine, a β
Journal of agricultural and food chemistry (2017-04-30)
Ying-Heng Chen, Chia-Ying Yang, Chih Wen Cheng, Yi-Ying Lin, Su Lien Kuo, Ling-Wei Hsin
ABSTRACT
4-[2-(t-Butylamino)-1-hydroxyethyl]phenol (buctopamine, 4), a new β
Sigma-Aldrich
2-Hydroxyacetophenone, 98%