Merck
  • Deciphering the molecular basis of invasiveness in Sdhb-deficient cells.
Deciphering the molecular basis of invasiveness in Sdhb-deficient cells.
Oncotarget (2015-10-16)
Céline Loriot, Mélanie Domingues, Adeline Berger, Mélanie Menara, Maëva Ruel, Aurélie Morin, Luis-Jaime Castro-Vega, Éric Letouzé, Cosimo Martinelli, Alexis-Pierre Bemelmans, Lionel Larue, Anne-Paule Gimenez-Roqueplo, Judith Favier
ABSTRACT
Metastatic pheochromocytomas and paragangliomas (PPGL) are malignant neuroendocrine tumors frequently associated with germline mutations in the SDHB gene. SDHB-mutated PPGL display a hypermethylator phenotype associated with hallmarks of epithelial-to-mesenchymal transition (EMT). In the present study, we report the characterization of a unique model of Sdhb knockout in mouse chromaffin cells. Sdhb deficient cells exhibit a metastatic phenotype as highlighted by increased individual cell migration (characterized by faster motility and increased persistence) as well as high invasive and adhesion abilities. This phenotype is associated with the modulation of Twist1, Twist2, Tcf3, Snai1, N-cadherin or Krt19 expression, reflecting an EMT-like reprogramming of cells. Krt19 is epigenetically silenced in Sdhb-deficient cells and re-expressed after treatment by the demethylating agent decitabine. Krt19 rescue by lentiviral transduction in Sdhb-deficient cells and Krt19 inhibition by RNA interference in wild-type cells were performed. Both studies revealed the involvement of KRT19 in the invasive phenotype by modulating collective and individual migration and cell/extra-cellular matrix adhesion properties. These findings underline the role of hypermethylation and EMT in the in vitro acquisition of metastatic properties, following SDHB loss of function.
Sigma-Aldrich
Anti-SDHB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%