Merck
An improved smaller biotin ligase for BioID proximity labeling.
Molecular biology of the cell (2016-02-26)
Dae In Kim, Samuel C Jensen, Kyle A Noble, Birendra Kc, Kenneth H Roux, Khatereh Motamedchaboki, Kyle J Roux
ABSTRACT
The BioID method uses a promiscuous biotin ligase to detect protein-protein associations as well as proximate proteins in living cells. Here we report improvements to the BioID method centered on BioID2, a substantially smaller promiscuous biotin ligase. BioID2 enables more-selective targeting of fusion proteins, requires less biotin supplementation, and exhibits enhanced labeling of proximate proteins. Thus BioID2 improves the efficiency of screening for protein-protein associations. We also demonstrate that the biotinylation range of BioID2 can be considerably modulated using flexible linkers, thus enabling application-specific adjustment of the biotin-labeling radius.
Sigma-Aldrich
Anti-Chicken IgY (IgG) (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid