Merck
  • Identification of a novel inactivating mutation in Isocitrate Dehydrogenase 1 (IDH1-R314C) in a high grade astrocytoma.
Identification of a novel inactivating mutation in Isocitrate Dehydrogenase 1 (IDH1-R314C) in a high grade astrocytoma.
Scientific reports (2016-07-28)
Sanne A M van Lith, Anna C Navis, Krissie Lenting, Kiek Verrijp, Jan T G Schepens, Wiljan J A J Hendriks, Nil A Schubert, Hanka Venselaar, Ron A Wevers, Arno van Rooij, Pieter Wesseling, Remco J Molenaar, Cornelis J F van Noorden, Stefan Pusch, Bastiaan Tops, William P J Leenders
ABSTRACT
The majority of low-grade and secondary high-grade gliomas carry heterozygous hotspot mutations in cytosolic isocitrate dehydrogenase 1 (IDH1) or the mitochondrial variant IDH2. These mutations mostly involve Arg132 in IDH1, and Arg172 or Arg140 in IDH2. Whereas IDHs convert isocitrate to alpha-ketoglutarate (α-KG) with simultaneous reduction of NADP(+) to NADPH, these IDH mutants reduce α-KG to D-2-hydroxyglutarate (D-2-HG) while oxidizing NADPH. D-2-HG is a proposed oncometabolite, acting via competitive inhibition of α-KG-dependent enzymes that are involved in metabolism and epigenetic regulation. However, much less is known about the implications of the metabolic stress, imposed by decreased α-KG and NADPH production, for tumor biology. We here present a novel heterozygous IDH1 mutation, IDH1(R314C), which was identified by targeted next generation sequencing of a high grade glioma from which a mouse xenograft model and a cell line were generated. IDH1(R314C) lacks isocitrate-to-α-KG conversion activity due to reduced affinity for NADP(+), and differs from the IDH1(R132) mutants in that it does not produce D-2-HG. Because IDH1(R314C) is defective in producing α-KG and NADPH, without concomitant production of the D-2-HG, it represents a valuable tool to study the effects of IDH1-dysfunction on cellular metabolism in the absence of this oncometabolite.
Sigma-Aldrich
Anti-IDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.