Merck
  • New bis(azobenzocrown)s with dodecylmethylmalonyl linkers as ionophores for sodium selective potentiometric sensors.
New bis(azobenzocrown)s with dodecylmethylmalonyl linkers as ionophores for sodium selective potentiometric sensors.
Journal of inclusion phenomena and macrocyclic chemistry (2016-11-25)
Elżbieta Luboch, Maciej Jeszke, Mirosław Szarmach, Natalia Łukasik
ABSTRACT
Novel biscrowns
Product Description
Supelco
Sodium ionophore VI, Selectophore, function tested
Sigma-Aldrich
Sodium 4-vinylbenzenesulfonate, technical, ≥90% (T)
Supelco
Potassium tetrakis(4-chlorophenyl)borate, Selectophore, ≥98.0%