Merck
  • Increased fibroblast chymase production mediates procollagen autophagic digestion in volume overload.
Increased fibroblast chymase production mediates procollagen autophagic digestion in volume overload.
Journal of molecular and cellular cardiology (2016-01-26)
Lianwu Fu, Chih-Chang Wei, Pamela C Powell, Wayne E Bradley, Sarfaraz Ahmad, Carlos M Ferrario, James F Collawn, Louis J Dell'Italia
ABSTRACT
Previous work has identified mast cells as the major source of chymase largely associated with a profibrotic phenotype. We recently reported increased fibroblast autophagic procollagen degradation in a rat model of pure volume overload (VO). Here we demonstrate a connection between increased fibroblast chymase production and autophagic digestion of procollagen in the pure VO of aortocaval fistula (ACF) in the rat. Isolated LV fibroblasts taken from 4 and 12week ACF Sprague-Dawley rats have significant increases in chymase mRNA and chymase activity. Increased intracellular chymase protein is documented by immunocytochemistry in the ACF fibroblasts compared to cells obtained from age-matched sham rats. To implicate VO as a stimulus for chymase production, we show that isolated adult rat LV fibroblasts subjected to 24h of 20% cyclical stretch induces chymase mRNA and protein production. Exogenous chymase treatment of control isolated adult cardiac fibroblasts demonstrates that chymase is internalized through a dynamin-dependent mechanism. Chymase treatment leads to an increased formation of autophagic vacuoles, LC3-II production, autophagic flux, resulting in increased procollagen degradation. Chymase inhibitor treatment reduces cyclical stretch-induced autophagy in isolated cardiac fibroblasts, demonstrating chymase's role in autophagy induction. In a pure VO model, chymase produced in adult cardiac fibroblasts leads to autophagic degradation of newly synthesized intracellular procollagen I, suggesting a new role of chymase in extracellular matrix degradation.
Sigma-Aldrich
Chymase human, recombinant, expressed in Pichia pastoris
Sigma-Aldrich
Anti-LC3 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Vimentin Antibody, serum, Chemicon®