Merck
  • Nuclear factor κB inhibitor BAY 11-7082 suppresses oxidative stress induced by endothelin-1 (ET-1) in rat kidney.
Nuclear factor κB inhibitor BAY 11-7082 suppresses oxidative stress induced by endothelin-1 (ET-1) in rat kidney.
Postepy higieny i medycyny doswiadczalnej (Online) (2015-01-01)
Agata Kowalczyk, Michał Kołodziejczyk, Anna Gorąca
ABSTRACT
The aim of the study was to evaluate the effect of BAY 11-7082, an NF-κB inhibitor, on basal and ET-1-induced production of reactive oxygen species (ROS), TNF-α and p65 protein in rat kidney. The experimental animals were divided into five groups (n=7) receiving: 1) saline (control); 2 and 3) ET-1 in a dose of 3 μg/kg body weight (b.w.) or 12.5 μg/kg b.w.; 4) BAY 11-7082 (10 mg/kg b.w.); 5) BAY 11-7082 (10 mg/kg b.w.) and ET-1 (12.5 μg/kg b.w.), respectively. In kidney homogenates the concentration of thiobarbituric acid reactive substances (TBARS), H2O2, TNF-α, p65 protein and GSH/GSSG ratio were determined. ET-1 resulted in a dose-dependent increase in TBARS and hydrogen peroxide (H2O2) levels, and a decrease in GSH/GSSG ratio when compared to the controls. BAY 11-7082 administered 1 h before ET-1 administration at a dose of 12.5 μg/kg resulted in a decrease (P<0.001) in TBARS and H2O2 levels and an increase (P<0.001) in GSH/GSSG ratio compared to the ET-1 groups. The level of TNF-α was increased (P<0.001) in the presence of ET-1, while BAY 11-7082 reduced the TNF-α level (P<0.001). The rats receiving BAY 11-7082 showed a decrease in NF-κB p65 protein level in the nuclear fraction and an increase in the cytoplasmic fraction. The results suggest that BAY 11-7082 plays a protective role against ET-1 induced oxidative stress in kidney tissue. These actions of BAY 11-7082 may result from reduced activity of NF-κB signaling pathways. Inhibition of the NF-κB pathway may be a promising strategy for preventing the progression of kidney damage.
Sigma-Aldrich
5-Sulfosalicylic acid hydrate, 95%
Sigma-Aldrich
1,1,3,3-Tetramethoxypropane, 99%
Sigma-Aldrich
2-Vinylpyridine, 97%