Merck
  • A rapid and simple procedure for the determination of synephrine in dietary supplements by gas chromatography-mass spectrometry.
A rapid and simple procedure for the determination of synephrine in dietary supplements by gas chromatography-mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2006-05-16)
Emilia Marchei, Simona Pichini, Roberta Pacifici, Manuela Pellegrini, Piergiorgio Zuccaro
ABSTRACT
A simple and rapid procedure based on gas chromatography-mass spectrometry (GC-MS) is described for determination of synephrine, active principle of Citrus aurantium plant, in solid and liquid dietary supplements. After the addition of 3,4-methylenedioxypropylamphetamine as internal standard (I.S.), a liquid-liquid extraction procedure in alkaline conditions with chloroform/isopropanol (9:1, v/v) was applied to the samples prior to analysis. Chromatography was performed on a fused capillary column and synephrine and I.S., derivatized with pentafluoropropionic anhydride, were determined in the selected-ion-monitoring (SIM) mode. The method was validated in the range 0.1-50 microg/mg or microg/mL synephrine. Mean recovery ranged between 89.3% and 90.5% in both solid and liquid dietary supplements. The quantification limit was 0.1 microg/mg or microg/ml. The method was applied to analysis of various dietary supplements promoted for aiding weight control containing, among other constituents such as ephedrine alkaloids and methylxanthines, Citrus aurantium. Amount of synephrine present in such products ranged from 3.1 microg/mg solid product to 480.2 microg/mL liquid product.
Product Description
Supelco
Pentafluoropropionic anhydride, for GC derivatization, LiChropur, 99%
Pricing and availability is not currently available.