Merck
  • Ultraviolet-ozone-treated PEDOT:PSS as anode buffer layer for organic solar cells.
Ultraviolet-ozone-treated PEDOT:PSS as anode buffer layer for organic solar cells.
Nanoscale research letters (2012-08-21)
Zisheng Su, Lidan Wang, Yantao Li, Haifeng Zhao, Bei Chu, Wenlian Li
ABSTRACT
Ultraviolet-ozone-treated poly(3,4-ethylenedioxythiophene):poly(styrene sulfonate) (PEDOT:PSS)was used as the anode buffer layer in copper phthalocyanine (CuPc)/fullerene-based solar cells. The power conversion efficiency of the cells with appropriated UV-ozone treatment was found to increase about 20% compared to the reference cell. The improved performance is attributed to the increased work function of the PEDOT:PSS layer, which improves the contact condition between PEDOT:PSS and CuPc, hence increasing the extraction efficiency of the photogenerated holes and decreasing the recombination probability of holes and electrons in the active organic layers.
Sigma-Aldrich
Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate), 1.3 wt % dispersion in H2O