Merck
  • Aurora A contributes to p150(glued) phosphorylation and function during mitosis.
Aurora A contributes to p150(glued) phosphorylation and function during mitosis.
The Journal of cell biology (2010-05-19)
Pierre Romé, Emilie Montembault, Nathalie Franck, Aude Pascal, David M Glover, Régis Giet
ABSTRACT
Aurora A is a spindle pole-associated protein kinase required for mitotic spindle assembly and chromosome segregation. In this study, we show that Drosophila melanogaster aurora A phosphorylates the dynactin subunit p150(glued) on sites required for its association with the mitotic spindle. Dynactin strongly accumulates on microtubules during prophase but disappears as soon as the nuclear envelope breaks down, suggesting that its spindle localization is tightly regulated. If aurora A's function is compromised, dynactin and dynein become enriched on mitotic spindle microtubules. Phosphorylation sites are localized within the conserved microtubule-binding domain (MBD) of the p150(glued). Although wild-type p150(glued) binds weakly to spindle microtubules, a variant that can no longer be phosphorylated by aurora A remains associated with spindle microtubules and fails to rescue depletion of endogenous p150(glued). Our results suggest that aurora A kinase participates in vivo to the phosphoregulation of the p150(glued) MBD to limit the microtubule binding of the dynein-dynactin complex and thus regulates spindle assembly.
Sigma-Aldrich
Anti-alpha-Tubulin Antibody, tyrosinated, clone YL1/2, clone YL 1/2, from rat
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
PC53097-1G1 g
Ships in 14 days - from Aldrich Partner.
€240.20
PC53097-5G5 g
Ships in 14 days - from Aldrich Partner.
€639.50
To order products, please contact your local dealer.