Merck
  • Ku70 is essential for histone deacetylase inhibitor trichostatin A-induced apoptosis.
Ku70 is essential for histone deacetylase inhibitor trichostatin A-induced apoptosis.
Molecular medicine reports (2015-02-20)
Jin Meng, Feng Zhang, Xu-Tao Zhang, Tao Zhang, Yu-Hua Li, Lei Fan, Yang Sun, He-Long Zhang, Qi-Bing Mei
ABSTRACT
It was previously reported that the histone deacetylase inhibitor (HDACI) trichostatin A (TSA) induced B cell lymphoma 2 (Bcl-2)-associated X protein (Bax)-dependent apoptosis in colorectal cancer (CRC) cells. In addition, Ku70 has been identified as a regulator of apoptosis, the mechanism of which proceeds via interacting with Bax. The aim of the present study was to investigate the role of Ku70 in TSA-induced apoptosis in the CRC cell lines HCT116 and HT29. The results showed that TSA induced the acetylation of Ku70, which was found to be associated with increased apoptosis. In addition, TSA treatment promoted the release of Bax from its complex with Ku70. Bax was then detected to have translocated from the cytoplasm into the mitochondria, while cytochrome c was detected to have translocated from the mitochondria into the cytoplasm. Furthermore, knockdown of Ku70 using small interfering RNA decreased TSA-induced apoptosis as well as downregulated the expression of Bax. These effects were rescued through pre-treatment of cells with the proteasome inhibitor MG132. In conclusion, the results of the present study suggested that Ku70 acetylation mediated TSA-induced apoptosis in CRC cells. In addition, Ku70 was found to be indispensable in TSA-induced apoptosis due to its role in protecting Bax from proteosomal degradation.
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M