Merck
  • Molecular structure and spectroscopic characterization of ethyl 4-aminobenzoate with experimental techniques and DFT quantum chemical calculations.
Molecular structure and spectroscopic characterization of ethyl 4-aminobenzoate with experimental techniques and DFT quantum chemical calculations.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-05-15)
S Muthu, E Isac Paulraj
ABSTRACT
The FT-IR and FT-Raman spectra of ethyl 4-aminobenzoate (EAB) in the solid phase were recorded. The fundamental vibrational wavenumbers, intensities of vibrational bands and the optimized geometrical parameters of the compound were evaluated using DFT (B3LYP) method with 6-311++G(d,p) basis set. The stable geometry of the compound was determined from the potential energy surface scan. Complete vibrational assignments and Natural Bond Orbital (NBO) analysis for the title compound were carried out. The assignments of the vibrational spectra were carried out with the help of normal co-ordinate analysis (NCA) following the Scaled Quantum Mechanical Force Field (SQMFF) methodology. The molecule orbital contributions were studied by using the total (TDOS), partial (PDOS), and overlap population (OPDOS) density of states. UV-visible spectrum of the compound was recorded and the electronic properties, such as HOMO and LUMO energies were performed by time-dependent DFT (TD-DFT) approach. Mulliken population analysis on atomic charges were also calculated. Besides, molecular electrostatic potential (MEP) and thermodynamic properties were performed.
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Pricing and availability is not currently available.