Merck
  • Stable TEMPO and ABNO Catalyst Solutions for User-Friendly (bpy)Cu/Nitroxyl-Catalyzed Aerobic Alcohol Oxidation.
Stable TEMPO and ABNO Catalyst Solutions for User-Friendly (bpy)Cu/Nitroxyl-Catalyzed Aerobic Alcohol Oxidation.
The Journal of organic chemistry (2015-10-13)
Janelle E Steves, Shannon S Stahl
ABSTRACT
Two solutions, one consisting of bpy/TEMPO/NMI and the other bpy/ABNO/NMI (bpy =2,2'-bipyridyl; TEMPO = 2,2,6,6-tetramethylpiperidine N-oxyl, ABNO = 9-azabicyclo[3.3.1]nonane N-oxyl; NMI = N-methylimidazole), in acetonitrile are shown to have good long-term stability (ā‰„1 year) under air at 5 Ā°C. The solutions may be combined in appropriate quantities with commercially available [Cu(MeCN)4]OTf to provide a convenient catalyst system for the aerobic oxidation of primary and secondary alcohols.
Sigma-Aldrich
Stahl Aerobic Oxidation TEMPO solution, 0.2 M in acetonitrile, Solution for Oxidation of Primary Alcohols
Sigma-Aldrich
Stahl Aerobic Oxidation ABNO solution, 0.04M ABNO in Acetonitrile, Solution for Oxidation of Primary and Secondary Alcohols