Merck
  • Crystalline desoxyribonuclease; isolation and general properties; spectrophotometric method for the measurement of desoxyribonuclease activity.
Crystalline desoxyribonuclease; isolation and general properties; spectrophotometric method for the measurement of desoxyribonuclease activity.
The Journal of general physiology (1950-03-01)
M KUNITZ
ABSTRACT
A crystalline enzyme capable of digesting thymus nucleic acid (desoxyribonucleic acid) has been isolated from fresh beef pancreas. The enzyme called "desoxyribonuclease" is a protein of the albumin type. Its molecular weight is about 60,000 and its isoelectric point is near pH 5.0. It contains about 8 per cent tyrosine and 2 per cent tryptophane. It is readily denatured by heat. The denaturation is reversible if heated in dilute acid at pH about 3.0. The digestion of thymus nucleic acid by crystalline desoxyribonuclease is accompanied by a gradual increase in the specific absorption of ultraviolet light by the acid. The spectrophotometric measurement of the rate of increase in the light absorption can be conveniently used as a general method for estimating desoxyribonuclease activity. Details are given of the method for isolation of crystalline desoxyribonuclease and of the spectrophotometric procedure for the measurement of desoxyribonuclease activity.
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Pricing and availability is not currently available.