Merck
  • Therapeutic potential of resveratrol in obesity and type 2 diabetes: new avenues for health benefits?
Therapeutic potential of resveratrol in obesity and type 2 diabetes: new avenues for health benefits?
Annals of the New York Academy of Sciences (2013-07-17)
Silvie Timmers, Matthijs K C Hesselink, Patrick Schrauwen
ABSTRACT
The number of people suffering from metabolic disorders is dramatically increasing worldwide. The need for new therapeutic strategies to combat this growing epidemic of metabolic diseases is therefore also increasing. In 2003, resveratrol was discovered to be a small molecule activator of sirtuin 1 (SIRT1), an important molecular target regulating cellular energy metabolism and mitochondrial homeostasis. Rodent studies have clearly demonstrated the potential of resveratrol to improve various metabolic health parameters. To date, however, only limited clinical data are available that have systematically examined the health benefits of resveratrol in metabolically challenged humans. This short review will give an overview of the currently available clinical studies examining the effects of resveratrol on obesity and type 2 diabetes from a human perspective.
Product Description
Resveratrol, European Pharmacopoeia (EP) Reference Standard
Supelco
Resveratrol, analytical standard
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)