Merck
  • Applications of functionalized and nanoparticle-modified nanocrystalline cellulose.
Applications of functionalized and nanoparticle-modified nanocrystalline cellulose.
Trends in biotechnology (2012-03-13)
Edmond Lam, Keith B Male, Jonathan H Chong, Alfred C W Leung, John H T Luong
ABSTRACT
Nanocrystalline cellulose (NCC), a rod-shaped nanoscale material with exceptional strength and physicochemical properties, can be prepared from inexpensive renewable biomass. Besides its potential use as a reinforcing agent for industrial biocomposites, pristine NCC exhibits low toxicity and poses no serious environmental concerns, providing impetus for its use in bioapplications. Here, we review recent developments in the use of modified NCC for emerging bioapplications, specifically enzyme immobilization, antimicrobial and medical materials, green catalysis, biosensing and controlled drug delivery. We focus on the modification of NCC with chemical functionalities and inorganic nanoparticles, reviewing practical considerations such as reusability, toxicity and scale-up capability.
Product Description
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
α-Cellulose, powder
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, acid washed, powder, for column chromatography