Merck
[Evidence-based therapeutic drug monitoring for nevirapine].
Therapie (2011-08-09)
Patrice Muret, Sarah Piedoux, Caroline Solas, Sylvie Quaranta
ABSTRACT
Nevirapine, a HIV non nucleosidic reverse transcriptase inhibitor, displays an inter-individual variability in its pharmacokinetics parameters, related to its hepatic metabolism. Based on literature, is the nevirapine therapeutic drug monitoring relevant? In naïve and pre-treated HIV infected patients, the probability of achieving and maintaining an undetectable HIV viral load was significantly associated with a nevirapine plasma trough concentration (C(trough)) > 4 000 ng/mL. The probability of virologic failure was significantly associated with a C(trough) < 3 000 ng/mL. Concerning the exposure-toxicity relationship, the emergence of hepatotoxicity was more frequently associated with high C(trough), especially in case of HCV coinfection. Non-randomized studies have reported the interest of nevirapine therapeutic drug monitoring to optimize the virologic response and, to a lesser extent, to prevent hepatotoxicity. Therefore, the level of evidence of the interest of nevirapine therapeutic drug monitoring is "recommended".
Product Description
Nevirapine (anhydrous), European Pharmacopoeia (EP) Reference Standard
Nevirapine for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nevirapine