Merck
Pirfenidone: in idiopathic pulmonary fibrosis.
Drugs (2011-09-10)
Natalie J Carter
ABSTRACT
Pirfenidone is an orally administered pyridine that has orphan designation for the treatment of mild to moderate idiopathic pulmonary fibrosis (IPF) in the EU. Pirfenidone 2403 mg/day for 72 weeks administered to patients with IPF was associated with a significantly lower mean decline in the percent predicted forced vital capacity than placebo (primary endpoint) according to data from one of two randomized, double-blind, multinational trials (studies 004 and 006 [also known as the CAPACITY trials]), and data from a pooled analysis of both trials. In another randomized, double-blind, multicentre Japanese trial, the adjusted mean in the change in vital capacity from baseline to week 52 was significantly lower in patients with IPF who received pirfenidone 1800 mg/day (considered to be comparable to the 2403 mg/day dose in studies 004 and 006 on a weight-normalized basis) than in those who received placebo (primary endpoint). Pirfenidone had an acceptable tolerability profile in clinical trials, with most adverse events being mild to moderate in severity.
Sigma-Aldrich
Pirfenidone, ≥97% (HPLC)