Merck
  • Hydroxyurea for sickle cell disease: a systematic review for efficacy and toxicity in children.
Hydroxyurea for sickle cell disease: a systematic review for efficacy and toxicity in children.
Pediatrics (2008-12-03)
John J Strouse, Sophie Lanzkron, Mary Catherine Beach, Carlton Haywood, Haeseong Park, Catherine Witkop, Renee F Wilson, Eric B Bass, Jodi B Segal
ABSTRACT
Hydroxyurea is the only approved medication for the treatment of sickle cell disease in adults; there are no approved drugs for children. Our goal was to synthesize the published literature on the efficacy, effectiveness, and toxicity of hydroxyurea in children with sickle cell disease. Medline, Embase, TOXLine, and the Cumulative Index to Nursing and Allied Health Literature through June 2007 were used as data sources. We selected randomized trials, observational studies, and case reports (English language only) that evaluated the efficacy and toxicity of hydroxyurea in children with sickle cell disease. Two reviewers abstracted data sequentially on study design, patient characteristics, and outcomes and assessed study quality independently. We included 26 articles describing 1 randomized, controlled trial, 22 observational studies (11 with overlapping participants), and 3 case reports. Almost all study participants had sickle cell anemia. Fetal hemoglobin levels increased from 5%-10% to 15%-20% on hydroxyurea. Hemoglobin concentration increased modestly (approximately 1 g/L) but significantly across studies. The rate of hospitalization decreased in the single randomized, controlled trial and 5 observational studies by 56% to 87%, whereas the frequency of pain crisis decreased in 3 of 4 pediatric studies. New and recurrent neurologic events were decreased in 3 observational studies of hydroxyurea compared with historical controls. Common adverse events were reversible mild-to-moderate neutropenia, mild thrombocytopenia, severe anemia, rash or nail changes (10%), and headache (5%). Severe adverse events were rare and not clearly attributable to hydroxyurea. Hydroxyurea reduces hospitalization and increases total and fetal hemoglobin levels in children with severe sickle cell anemia. There was inadequate evidence to assess the efficacy of hydroxyurea in other groups. The small number of children in long-term studies limits conclusions about late toxicities.
Product Description
Hydroxycarbamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydroxyurea, 98%, powder