Merck
[The use of gliclazide in individualized sulfonylurea therapy].
Orvosi hetilap (2014-04-01)
Gábor Winkler
ABSTRACT
In addition to the common blood glucose lowering effect, sulfonylurea compounds are different in many aspects from each other. Based on earlier findings the second generation gliclazide has special advantages within this group. Although the number of experimental and clinical observations on gliclazide is continuously increasing, these novel findings are not in the focus anymore due to the appearance of new antidiabetics. This article reviews recent experimental (effect on receptors, the absence of Epac2 activation, antioxidant properties, possible incentive of factors participating in beta-cell differentiation) and pharmacogenomic data, and compares them with clinical observations obtained from gliclazide treatment (hypoglycemias, parameters of cardiovascular outcome). The data underline the advantages of gliclazide, the highly pancreas-selective nature, preservation of the ischemic precondition, favourable hemodynamic properties and potential reduction of the beta-cell loss as compared to other compounds of the group. However, gliclazide is not free from disadvantages characteristic to sulfonylureas in general (blood glucose independent insulin stimulation, beta-cell depletion). Comparing gliclazide with other derivatives of the group, the above data indicate individual benefits for the application when sulfonylurea compound is the drug of choice.
Product Description
Supelco
Gliclazide, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Gliclazide, powder, ≥98%
Pricing and availability is not currently available.
Gliclazide, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.