Merck
  • Room-temperature Pd-catalyzed amidation of aryl bromides using tert-butyl carbamate.
Room-temperature Pd-catalyzed amidation of aryl bromides using tert-butyl carbamate.
The Journal of organic chemistry (2009-06-13)
Swapna Bhagwanth, Alex G Waterson, George M Adjabeng, Keith R Hornberger
ABSTRACT
The scope of Pd-catalyzed synthesis of N-Boc-protected anilines from aryl bromides and commercially available tert-butyl carbamate is described. For the first time, this process can be conducted at room temperature (17-22 degrees C) using a combination of Pd(2)dba(3).CHCl(3) and a monodentate ligand, tert-butyl X-Phos. Use of sodium tert-butoxide is crucial to the success of the reaction, which proceeds in 43-83% yield.
Sigma-Aldrich
tert-Butyl carbamate, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
1 mg
1 Available to ship on January 08, 2025
Details...
MXP 1,557.00
To order products, please contact your local dealer.