Merck
  • Vitamin K status in human tissues: tissue-specific accumulation of phylloquinone and menaquinone-4.
Vitamin K status in human tissues: tissue-specific accumulation of phylloquinone and menaquinone-4.
The British journal of nutrition (1996-01-01)
H H Thijssen, M J Drittij-Reijnders
ABSTRACT
We measured the vitamin K status in postmortem human tissues (brain, heart, kidney, liver, lung, pancreas) to see if there is a tissue-specific distribution pattern. Phylloquinone (K1) was recovered in all tissues with relatively high levels in liver, heart and pancreas (medians, 10.6 (4.8), 9.3 (4.2), 28.4 (12.8) pmol(ng)/g wet weight tissue); low levels (< 2 pmol/g) were found in brain, kidney and lung. Menaquinone-4 (MK-4) was recovered from most of the tissues; its levels exceeded the K1 levels in brain and kidney (median, 2.8 ng/g) and equalled K1 in pancreas. Liver, heart and lung were low in MK-4. The higher menaquinones, MK-6-11, were recovered in the liver samples (n 6), traces of MK-6-9 were found in some of the heart and pancreas samples. The results show that in man there are tissue-specific, vitamin-K distribution patterns comparable to those in the rat. Furthermore, the accumulation of vitamin K in heart, brain and pancreas suggests a hitherto unrecognized physiological function of this vitamin.
Product Description
Supelco
Menadione (K3), analytical standard
Sigma-Aldrich
Menadione, meets USP testing specifications
Menadione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Menadione, crystalline