Merck
  • Merbarone inhibits the catalytic activity of human topoisomerase IIalpha by blocking DNA cleavage.
Merbarone inhibits the catalytic activity of human topoisomerase IIalpha by blocking DNA cleavage.
The Journal of biological chemistry (1998-07-04)
J M Fortune, N Osheroff
ABSTRACT
Merbarone is a catalytic inhibitor of topoisomerase II that is in clinical trials as an anticancer agent. Despite the potential therapeutic value of this drug, the mechanism by which it blocks topoisomerase II activity has not been delineated. Therefore, to determine the mechanistic basis for the inhibitory action of merbarone, the effects of this drug on individual steps of the catalytic cycle of human topoisomerase IIalpha were assessed. Concentrations of merbarone that inhibited catalytic activity >/=80% had no effect on either enzyme.DNA binding or ATP hydrolysis. In contrast, the drug was a potent inhibitor of enzyme-mediated DNA scission (in the absence or presence of ATP), and the inhibitory profiles of merbarone for DNA cleavage and relaxation were similar. These data indicate that merbarone acts primarily by blocking topoisomerase II-mediated DNA cleavage. Merbarone inhibited DNA scission in a global (rather than site-specific) fashion but did not appear to intercalate into DNA or bind in the minor groove. Since the drug competed with etoposide (a cleavage-enhancing agent that binds directly to topoisomerase II), it is proposed that merbarone exerts its inhibitory effects through interactions with the enzyme and that the drug shares an interaction domain on topoisomerase II with cleavage-enhancing agents.
Sigma-Aldrich
Merbarone, ≥98% (HPLC), solid