Merck
  • Colorimetric detection of achiral anions and chiral carboxylates by a chiral thiourea-phthalimide dyad.
Colorimetric detection of achiral anions and chiral carboxylates by a chiral thiourea-phthalimide dyad.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2010-09-08)
Axel G Griesbeck, Sebastian Hanft, Yrene Díaz Miara
ABSTRACT
The chiral chemosensor 1, based on a thiourea-activated phthalimide, is available by four reaction steps from 4-nitrophthalimide. 1 detects fluoride, chloride, acetate, and dihydrogen phosphate anions by changes in UV-vis absorption. Fluoride in excess induces deprotonation whereas the other anions show only complex formation in the ground state. (1)H-NMR studies confirm the formation of these H-bonded complexes and the fluoride-induced receptor deprotonation in the recognition process. Moderate chiral recognition was observed for sodium D/L-lactate with K(ass)(D)/K(ass)(L) = 1.93.
Sigma-Aldrich
4-Nitrophthalimide, 98%
Pricing and availability is not currently available.