Merck
  • Macular hole surgery and cataract extraction: combined vs consecutive surgery.
Macular hole surgery and cataract extraction: combined vs consecutive surgery.
American journal of ophthalmology (2010-07-10)
Aurore Muselier, Brice Dugas, Xavier Burelle, Marie Passemard, Isabelle Hubert, Bénigne Mathieu, Jean Paul Berrod, Alain M Bron, Catherine Creuzot-Garcher
ABSTRACT
To compare the functional and the anatomic outcomes of a combined surgery and consecutive surgery for macular hole and cataract extraction. Multicenter, retrospective, comparative case series. One hundred twenty patients (120 eyes) with an idiopathic macular hole and cataract were operated on in 1 or 2 sessions in 2 academic centers, Dijon University Hospital and Nancy University Hospital. Combined surgery (n = 64) and consecutive surgery (n = 56) were performed between 2006 and 2007. All patients underwent pars plana vitrectomy with internal limiting membrane peeling and gas tamponade. Cataract extraction was performed with phacoemulsification followed by a posterior chamber intraocular lens implantation. The main outcome measures were near and far visual acuity at 6 and 12 months, and the rate of closure of macular hole evaluated with optical coherence tomography. After a 12-month follow-up, the postoperative best-corrected visual acuities significantly improved in both the combined and the consecutive surgery groups (near and far vision in both groups, P < .0001). However the improvement of far visual acuity was not significant in the consecutive surgery group at 6 months (P = .06) while such an improvement was observed in the combined surgery group (P < .0001). The rates of closure, 100% and 96% in the combined and the consecutive groups respectively, and the complications did not differ significantly between groups. Both combined and consecutive surgeries are safe and effective methods to treat macular hole and cataract with equivalent functional and anatomic results in both procedures. However, combined surgery shortened the delay for visual recovery.
Sigma-Aldrich
Hexafluoroethane, ≥98%