Merck
Metabolism of dimethylterephthalate by Aspergillus niger.
Biodegradation (1995-01-01)
S H Ganji, C S Karigar, B G Pujar
ABSTRACT
Aspergillus niger (AG-1) metabolized dimethylterephthalate through monomethylterephthalate, terephthalate and protocatechuate. Degradation of dimethylterephthalate was followed by extraction of residual dimethylterephthalate from the spent medium. The quantitative UV analysis showed that 58% of the dimethylterephthalate supplement was taken up in 144 h. The metabolites were isolated from resting cell cultures. Thin layer chromatography analysis of the extract revealed the presence of two intermediates, monomethylterephthalate and terephthalate. Use of an inhibitor in resting cell culture experiment demonstrated the accumulation of protocatechuate. The time course of protocatechuate accumulation was also studied. Metabolites were identified by employing various physicochemical methods. Enzyme studies using cell-free extracts exhibited dimethylterephthalate esterase and protocatechuate dioxygenase activities. Protocatechuate was oxidized by the meta cleavage pathway. A tentative pathway for the degradation of DMTP has been proposed in A. niger.
Sigma-Aldrich
Dimethyl terephthalate, ReagentPlus®, ≥99%
Supelco
Dimethyl terephthalate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl terephthalate, purum, ≥99.0% (GC)