Merck
Rosin: a naturally derived excipient in drug delivery systems.
Polimery w medycynie (2013-07-03)
Shobhit Kumar, Satish Kumar Gupta
ABSTRACT
Natural polymers are primarily attractive because they are biodegradable, inexpensive, and readily available. The most important benefit of natural polymers is that they are capable for chemical modifications. One such biopolymer, rosin, and its derivatives have been pharmaceutically evaluated as microencapsulating materials, film forming agent and as binding agent in formulation of tablets. They are also employed in formulation of chewing gum bases and cosmetics. This review article provides an overview of pharmaceutical use of rosin and its derivatives as excipient in dosage forms as well as novel drug delivery systems.
Sigma-Aldrich
Gum rosin, CP
Sigma-Aldrich
Gum rosin, natural resin