Merck
  • Enantioselective synthesis of atropisomeric benzamides through peptide-catalyzed bromination.
Enantioselective synthesis of atropisomeric benzamides through peptide-catalyzed bromination.
Journal of the American Chemical Society (2013-02-16)
Kimberly T Barrett, Scott J Miller
ABSTRACT
We report the enantioselective synthesis of atropisomeric benzamides employing catalytic electrophilic aromatic substitution reactions involving bromination. The catalyst is a simple tetrapeptide bearing a tertiary amine that may function as a Brønsted base. A series of tri- and dibrominations were accomplished for a range of compounds bearing differential substitution patterns. Tertiary benzamides represent appropriate substrates for the reaction since they exhibit sufficiently high barriers to racemization after ortho functionalization. Mechanism-driven experiments provided some insight into the basis for selectivity. Examination of the observed products at low conversion suggested that the initial catalytic bromination may be regioselective and stereochemistry-determining. A complex between the catalyst and substrate was observed by NMR spectroscopy, revealing a specific association. Finally, the products of these reactions may be subjected to regioselective metal-halogen exchange and trapping with I(2), setting the stage for utility.
Sigma-Aldrich
Bromine water, CP
Sigma-Aldrich
Bromine, ≥99.99% trace metals basis
Sigma-Aldrich
Bromine, SAJ first grade, ≥97.0%
Sigma-Aldrich
Bromine, JIS special grade, ≥99.0%
Sigma-Aldrich
Bromine, reagent grade
Sigma-Aldrich
Bromine, ACS reagent, ≥99.5%