Merck
Functionalized coronenes: synthesis, solid structure, and properties.
The Journal of organic chemistry (2012-11-29)
Di Wu, Hua Zhang, Jinhua Liang, Haojie Ge, Chunyan Chi, Jishan Wu, Sheng Hua Liu, Jun Yin
ABSTRACT
The construction of coronenes using simple building blocks is a challenging task. In this work, triphenylene was used as a building block to construct functionalized coronenes, and their solid structures and optoelectronic properties were investigated. The single crystal structures showed that coronenes have different packing motifs. Their good solubility and photostability make them potential solution-processable candidates for organic devices.
Sigma-Aldrich
Coronene, 97%
Sigma-Aldrich
Coronene, purified by sublimation, 99%
Coronene, BCR®, certified reference material