Merck
  • Improvement of a puromycin-linker to extend the selection target varieties in cDNA display method.
Improvement of a puromycin-linker to extend the selection target varieties in cDNA display method.
Journal of biotechnology (2012-09-25)
Shingo Ueno, Shinnosuke Kimura, Takanori Ichiki, Naoto Nemoto
ABSTRACT
cDNA display using a puromycin-linker to covalently bridge a protein and its coding cDNA is a stable and efficient in vitro protein selection method. The optimal design of the often-used puromycin-linker is vital for effective selection. In this report, an improved puromycin-linker containing deoxyinosine bases as cleavage sites, which are recognized by endonuclease V, was introduced to extend the variety of the selection targets to molecules such as RNA. The introduction of this linker enables efficient in vitro protein selection without contamination from RNase T1, which is used for the conventional linker containing ribonucleotide G bases. In addition, mRNA-protein fusion efficiency was found to not depend on the length of the flexible poly (ethylene glycol) (PEG) region of the linker. These findings will allow practical and easy-to-use in vitro protein selection by cDNA display.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture