Merck
[Effect of shengbanfang on CYP3A1 activities of Sprague-Dawley rat].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2012-07-25)
Tao Liu, Duo Deng, Zi-chao Lin, Ying Pan, Wen-jing Zhou, Wang Zhou, Hong-bing Huang
ABSTRACT
To study the influences of Shengbanfang on CYP3A1 activities of SD rat and provide suggestions for drug combinations. 25 male SD rats were devided into 5 groups randomly,and treated with saline( NS, ig, 10 mg/(kg/d) ,qd,14 d), dexamethasone (DEX, ig, 100 mg/(kg x d), qd, 3 d), high dose of Shengbanfang (HD, ig, 8.645 g/kg, bid, 14 d), middle dose of Shengbanfang (MD, ig, 4.322 g/kg, bid, 14 d) and low dose of Shengbanfang (LD, ig, 2.161 g/kg, bid, 14 d), respectively. The HPLC method was established and validated to determine the productive velocity of 6beta-hydroxytestosterone and measure the activity of CYP3Al. Under the optimized incubation conditions, the productive rates of 6beta-hydroxytestosterone of HD, MD, LD, NS and DEX, groups were (55.82 +/- 5.97), (65.10 +/- 6.83), (60.89 +/- 6.53), (62.17 +/- 6.55), (126.73 +/- 15.40) micromol/(L x mg pro x min). There were significant differences between Shengbanfang groups compared with dexamethasone group, but there was no significant difference between Shengbanfang groups and the control group (NS). Shengbanfang has no induce effect on the enzymic activity of CYP3Al in SD rats.
Sigma-Aldrich
6β-Hydroxytestosterone, ≥97% (HPLC)
Supelco
6β-Hydroxytestosterone solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®