Merck
  • Sulfinyl-mediated stereoselective Overman rearrangements and Diels-Alder cycloadditions.
Sulfinyl-mediated stereoselective Overman rearrangements and Diels-Alder cycloadditions.
Organic letters (2012-05-29)
Roberto Fernández de la Pradilla, Ignacio Colomer, Alma Viso
ABSTRACT
The Overman rearrangement of allylic sulfinyl trichloroacetimidates affords sulfinyl trichloroacetamides with high stereoselectivity and excellent yields. Bis-allylic substrates lead to amido 2-sulfinyl butadiene derivatives in excellent yields, with total chemo- and diastereoselectivity. The Diels-Alder cycloaddition of related dienes is controlled by the sulfoxide moiety.
Sigma-Aldrich
Safrole, ≥97%
Supelco
Safrole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland