Merck
  • Reevaluation of copper(I) affinity for amyloid-β peptides by competition with ferrozine--an unusual copper(I) indicator.
Reevaluation of copper(I) affinity for amyloid-β peptides by competition with ferrozine--an unusual copper(I) indicator.
Chemistry (Weinheim an der Bergstrasse, Germany) (2011-12-23)
Bruno Alies, Bertrand Badei, Peter Faller, Christelle Hureau
ABSTRACT
The association constant of ferrozine (5,6-diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4''-disulfonic acid) with Cu(I) to form the chromophoric [Cu(I)(Fz)(2)](3-) complex was determined by UV/Vis titration experiments in Hepes buffer (0.1 M, pH 7.4). An association constant close to 10(12) M(-2), which is significantly weaker than those of the well-known, water-soluble, Cu(I) chelators bicinchoninic acid and 2,9-dimethyl-4,7-diphenyl-1,10-phenantroline disulfonic acid, was found. The [Cu(I)(Fz)(2)](3-) chromophore was used in UV/Vis competition experiments to determine Cu(I) binding affinity for the amyloid-β peptide involved in Alzheimer's disease and for a series of pertinent mutants. An association constant of approximately 10(7) M(-1) was found; this is much weaker than that reported for dithiothreitol and confirms that imidazoles are harder ligands than thiolates. Each His mutation (H6A, H13A, and H14A) impacts the peptide affinity for Cu(I). The native human amyloid-β peptide was found to be a fourfold-stronger Cu(I) ligand than the murine peptide, which differs by three point mutations (R5G, Y10F, and H13R) from the human one.
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p′-disulfonic acid monosodium salt hydrate, 97%
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, BioXtra
Sigma-Aldrich
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, Indicator ligand
Supelco
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-4′,4′′-disulfonic acid sodium salt, for spectrophotometric det. of Fe, 97.0%