- Influence of treatment with intranasal corticosteroids on the nasal mucosa, weight, and corticosteroid concentration in rats.
Influence of treatment with intranasal corticosteroids on the nasal mucosa, weight, and corticosteroid concentration in rats.
The effect of intranasal corticosteroids on the nasal epithelium mucosa is an important parameter of treatment safety. This study was designed to examine whether treatment with topical corticosteroids in patients with allergic rhinitis causes atrophic nasal mucosal changes, when compared with systemic corticosteroids, in rats. Male Wistar rats were treated daily during 7 weeks with topical administration with 10 microliters of normal saline (control group), 10 microliters of mometasone furoate group, 10 microliters of triamcinolone acetonide (T group), and 8 mg/kg of daily subcutaneous injections of methylprednisolone sodium succinate (MP group). Body weight was evaluated weekly. At the end of the treatment, rats were killed by decapitation to collect blood for determination of corticosterone levels and nasal cavities were prepared for histological descriptive analyses. Treatment with T and MP decreased body weight. Plasma corticosterone concentration was significantly reduced by MP treatment and presented a clear tendency to decrease after T treatment. Histological changes observed in group T included ripples, cell vacuolization, increase in the number of nuclei, and decrease in the number of cilia in the epithelial cells. Growth and corticosterone concentration were impaired by T and MP at the same proportion, suggesting a role of this hormone in body gain. With the exception of T, intranasal or systemic treatment with the corticosteroids evaluated in this study did not affect nasal mucosa.