Merck
  • Organic and inorganic selenium: II. Transfer efficiency from ewes to lambs.
Organic and inorganic selenium: II. Transfer efficiency from ewes to lambs.
Journal of animal science (2011-10-04)
W C Stewart, G Bobe, W R Vorachek, G J Pirelli, W D Mosher, T Nichols, R J Van Saun, N E Forsberg, J A Hall
ABSTRACT
Adequate Se transfer from ewes to lambs is important to prevent Se-deficiency diseases. To evaluate how different chemical forms of Se administered at comparative dosages to mature ewes affect Se status of their lambs, 240 ewes were divided into 8 treatment groups (n = 30 each) and drenched weekly (at an amount equal to their summed daily intake) with no-Se (controls); at recommended amounts (4.9 mg of Se/wk) with inorganic Na-selenite, inorganic Na-selenate, or organic Se-yeast; or at supranutritional amounts (14.7 and 24.5 mg of Se/wk) with Na-selenite or Se-yeast for 1 yr. Weekly drenching of Se was effective at increasing (P < 0.002) Se concentrations in ewe colostrum and milk at 30 d of lactation and in improving (P < 0.001) the Se status of lambs (whole-blood and serum-Se concentrations at birth, and skeletal-muscle Se concentrations at 14 d of age). Selenium concentrations in lacteal secretions were greater in ewes drenched with Se-yeast (colostrum: 374, 436, and 982 ng/mL at 4.9, 14.7, and 24.5 mg of Se/wk, respectively; milk: 26, 39, 64 ng/mL) compared with ewes drenched with Na-selenite (colostrum: 204, 334, 428 ng/mL; milk: 16, 21, 24 ng/mL), and were also greater (P < 0.001) in their lambs. Selenium concentrations continued to increase (P < 0.001) in lamb whole blood (558 and 695 ng/mL at 14.7 and 24.5 mg of Se/wk, respectively), serum (126, 183 ng/mL), and skeletal muscle (991, 1,696 ng/mL) with supranutritional concentrations of Se-yeast, whereas Se concentrations did not differ in whole blood (304, 332 ng/mL), serum (77, 85 ng/mL), or skeletal muscle (442, 482 ng/mg) of lambs from ewes drenched with 14.7 or 24.5 mg of Se/wk of Na-selenite. We conclude that weekly oral drenching of ewes during gestation and lactation with organic Se-yeast results in a more efficient transfer of Se (over a wide range of supplementation rates) from ewe to lamb than does inorganic Na-selenite.
Sigma-Aldrich
Sodium selenate, ≥95% (elemental analysis)
Sigma-Aldrich
Sodium selenate, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Sodium selenate, BioXtra