Merck
  • A cytotoxic type-2 ribosome inactivating protein (from leafless mistletoe) lacking sugar binding activity.
A cytotoxic type-2 ribosome inactivating protein (from leafless mistletoe) lacking sugar binding activity.
International journal of biological macromolecules (2011-09-21)
Mrinal Kumar Das, Radhey Shyam Sharma, Vandana Mishra
ABSTRACT
Articulatin-D, a 66 kDa ribosome inactivating protein (RIP) comprised of 29 kDa A-chain linked to 35 kDa B-chain, is purified from leafless mistletoe (Viscum articulatum) parasitic on Dalbergia sp. from Western Ghats (India). N-terminal sequence and LC-MS/MS analyses of A- and B-chain confirmed that articulatin-D is a type-2 RIP having high homology with other mistletoe lectins. Translation inhibition and diagnostic N-glycosidase activity of articulatin-D illustrate the presence of catalytically active A-chain. Its inability to: (i) bind to acid treated Sepharose CL-6B column, (ii) agglutinate trypsin-treated and untreated RBCs of human (A, B, O, AB), mice, rat, rabbit, buffalo, porcine, pigeon, cock, fish, sheep and goat even with 10mg/ml of purified articulatin-D, (iii) show change in circular dichroism spectra after addition of sugar to the native protein, (iv) bind to different sugars (galactose, lactose, gal-NAc, rhamnose, arabinose, fucose and mannose) immobilized on Sepharose 4B matrix, and (v) show change in enthalpy during titration with galactose confirm that the B-chain of articulatin-D lacks sugar binding activity. Despite this, articulatin-D is highly toxic as characterized with low IC(50) against different cancer cell lines (Jurkat: 0.31 ± 0.02 nM, MOLT-4: 0.51 ± 0.03 nM, U-937: 0.64 ± 0.07 nM, HL-60: 0.79 ± 0.11 nM, Raji: 1.45 ± 0.09 nM). Toxicity of RIPs has been ascribed to the absence/presence of B-chain with sugar binding activity. Identification of articulatin-D, the first cytotoxic RIP with B-chain lacking sugar binding activity opens new vistas in understanding cytotoxic action of RIPs.
Sigma-Aldrich
Sepharose CL-6B, Cross-linked