Merck
  • Development and application of a multimodal contrast agent for SPECT/CT hybrid imaging.
Development and application of a multimodal contrast agent for SPECT/CT hybrid imaging.
Bioconjugate chemistry (2011-08-20)
Jason M Criscione, Lawrence W Dobrucki, Zhen W Zhuang, Xenophon Papademetris, Michael Simons, Albert J Sinusas, Tarek M Fahmy
ABSTRACT
Hybrid or multimodality imaging is often applied in order to take advantage of the unique and complementary strengths of individual imaging modalities. This hybrid noninvasive imaging approach can provide critical information about anatomical structure in combination with physiological function or targeted molecular signals. While recent advances in software image fusion techniques and hybrid imaging systems have enabled efficient multimodal imaging, accessing the full potential of this technique requires development of a new toolbox of multimodal contrast agents that enhance the imaging process. Toward that goal, we report the development of a hybrid probe for both single photon emission computed tomography (SPECT) and X-ray computed tomography (CT) imaging that facilitates high-sensitivity SPECT and high spatial resolution CT imaging. In this work, we report the synthesis and evaluation of a novel intravascular, multimodal dendrimer-based contrast agent for use in preclinical SPECT/CT hybrid imaging systems. This multimodal agent offers a long intravascular residence time (t(1/2) = 43 min) and sufficient contrast-to-noise for effective serial intravascular and blood pool imaging with both SPECT and CT. The colocalization of the dendritic nuclear and X-ray contrasts offers the potential to facilitate image analysis and quantification by enabling correction for SPECT attenuation and partial volume errors at specified times with the higher resolution anatomic information provided by the circulating CT contrast. This may allow absolute quantification of intramyocardial blood volume and blood flow and may enable the ability to visualize active molecular targeting following clearance from the blood.
Sigma-Aldrich
2,3,5-Triiodobenzoic acid, BioReagent, suitable for plant cell culture, ≥97% (HPLC)
Sigma-Aldrich
2,3,5-Triiodobenzoic acid, 98%