Merck
  • Intraperitoneal photodynamic therapy mediated by a fullerene in a mouse model of abdominal dissemination of colon adenocarcinoma.
Intraperitoneal photodynamic therapy mediated by a fullerene in a mouse model of abdominal dissemination of colon adenocarcinoma.
Nanomedicine : nanotechnology, biology, and medicine (2011-06-08)
Pawel Mroz, Yumin Xia, Daisuke Asanuma, Aaron Konopko, Timur Zhiyentayev, Ying-Ying Huang, Sulbha K Sharma, Tianhong Dai, Usman J Khan, Tim Wharton, Michael R Hamblin
ABSTRACT
Functionalized fullerenes represent a new class of photosensitizer (PS) that is being investigated for photodynamic therapy (PDT) of various diseases, including cancer. We tested the hypothesis that fullerenes could be used to mediate PDT of intraperitoneal (IP) carcinomatosis in a mouse model. In humans this form of cancer responds poorly to standard treatment and manifests as a thin covering of tumor nodules on intestines and on other abdominal organs. We used a colon adenocarcinoma cell line (CT26) stably expressing luciferase to allow monitoring of IP tumor burden in BALB/c mice by noninvasive real-time optical imaging using a sensitive low-light camera. IP injection of a preparation of N-methylpyrrolidinium-fullerene formulated in Cremophor-EL micelles, followed by white-light illumination delivered through the peritoneal wall (after creation of a skin flap), produced a statistically significant reduction in bioluminescence and a survival advantage in mice. This team of investigators report on functionalized fullerenes, to be used as photosensitizer for photodynamic therapy and demonstrate the efficacy of this method in an intraperitoneal carcinomatosis mouse model.
Sigma-Aldrich
1-Methylpyrrolidine, ≥98.0% (GC)
Sigma-Aldrich
N-Methylpyrrolidine, 97%