Merck
  • Developmental competence of parthenogenetic mouse and human embryos after chemical or electrical activation.
Developmental competence of parthenogenetic mouse and human embryos after chemical or electrical activation.
Reproductive biomedicine online (2010-11-06)
Karen Versieren, Björn Heindryckx, Sylvie Lierman, Jan Gerris, Petra De Sutter
ABSTRACT
Parthenogenetic reconstruction is one major strategy to create patient-specific stem cells. The aim of this study was to find the best artificial activation protocol for parthenogenetic activation of mouse and human oocytes comparing different methods. In a first set of experiments, in-vivo matured mouse oocytes and human failed-fertilized, in-vitro and in-vivo matured oocytes were artificially activated by a chemical (ionomycin) or electrical stimulus. In a second set of experiments, a combination of activating agents (electrical pulses followed by ionomycin or SrCl(2)) was applied in an aim to improve developmental competence. All embryos were evaluated daily until day 6 after activation. Mouse blastocysts were differentially stained to evaluate blastocyst quality. For mouse oocytes and human failed-fertilized oocytes, blastocyst development was significantly higher after electrical activation (P<0.05). For human in-vitro and in-vivo matured oocytes, blastocyst formation was only obtained after electrical activation of in-vitro matured oocytes. After combining activating agents, no differences in development could be observed. In conclusion, this study revealed that for both mouse and human oocytes development to the blastocyst stage was significantly better after electrical activation compared with chemical activation. Combining activating agents had no further positive effect on developmental potential.
Sigma-Aldrich
Strontium chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Strontium chloride solution, 0.1 M
Sigma-Aldrich
Strontium chloride hexahydrate, ACS reagent, 99%
Sigma-Aldrich
Strontium chloride hexahydrate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Strontium chloride hexahydrate, 99.995% trace metals basis